Isopropylamine, 99%
Isopropylamine, 99%
Isopropylamine, 99%
Thermo Scientific Chemicals
Isopropylamine, 99%
CAS: 75-31-0 | C3H9N | 59.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
1 L
Packaging:
Glass bottle
Catalog number FSA148920250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialIsopropylamine
CAS75-31-0
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
May cause respiratory irritation.
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Extremely flammable liquid and vapor.
Health Hazard 3GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse