Methyl thioglycolate, 98%
Methyl thioglycolate, 98%
Methyl thioglycolate, 98%
Methyl thioglycolate, 98%
Thermo Scientific Chemicals
Methyl thioglycolate, 98%
CAS: 2365-48-2 | C3H6O2S | 106.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA148881000
Price (MYR)
277.00
EA
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
277.00
EA
Add to cart
Specifications
Chemical Name or MaterialMethyl Thioglycolate
CAS2365-48-2
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement:
Causes serious eye irritation.
Toxic if swallowed.
Causes skin irritation.
Flammable liquid and vapor.
Health Hazard 3GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.