4-Nitro-o-phenylenediamine, 98%
4-Nitro-o-phenylenediamine, 98%
4-Nitro-o-phenylenediamine, 98%
4-Nitro-o-phenylenediamine, 98%
Thermo Scientific Chemicals
4-Nitro-o-phenylenediamine, 98%
CAS: 99-56-9 | C6H7N3O2 | 153.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA148841000
Price (MYR)
833.00
EA
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
833.00
EA
Add to cart
Specifications
Chemical Name or Material4-Nitro-o-phenylenediamine
Name Note98%
CAS99-56-9
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning