Allylthiourea, 98%
Allylthiourea, 98%
Allylthiourea, 98%
Thermo Scientific Chemicals
Allylthiourea, 98%
CAS: 109-57-9 | C4H8N2S | 116.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Catalog number FSA148805000
Price (MYR)
2,176.00
BOT
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
2,176.00
BOT
Add to cart
Specifications
Chemical Name or MaterialAllylthiourea
Name Note98%
CAS109-57-9
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger