Hexachloroethane, 99%
Hexachloroethane, 99%
Hexachloroethane, 99%
Hexachloroethane, 99%
Thermo Scientific Chemicals
Hexachloroethane, 99%
CAS: 67-72-1 | C2Cl6 | 236.72 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
1 kg
2.5 kg
Packaging:
Plastic bottle
Catalog number FSA148541000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialHexachloroethane
CAS67-72-1
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Suspected of causing cancer.
Toxic to aquatic life with long lasting effects.
Health Hazard 3GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF IN
View more
C-X bond formation reagent
RUO – Research Use Only