Top Grain Cowhide Leather Glove Protectors
Top Grain Cowhide Leather Glove Protectors
Actual product may vary
Thermo Scientific Chemicals
Top Grain Cowhide Leather Glove Protectors
For use with Class 1-2 Rubber Insulating Novax™ Gloves. Leather protectors should only be used with mechanical protection for rubber insulating gloves. Not to be used for electrical protection per ASTM F696-06 (2011) Standards.
Have Questions?
Catalog NumberSize
FSA14840001010
Catalog number FSA148400010
Price (MYR)
321.00
BOT
Add to cart
Size:
10
Price (MYR)
321.00
BOT
Add to cart
  • Top grain cowhide leather construction provides comfort, durability, abrasion resistance, and breathability
  • Split leather gauntlet cuff for added forearm protection, and fast removal
  • Straight thumb increases comfort in closed-fisted work
  • Nylon strap with buckle for a secure, personal fit
  • Color coded hems for easy size identification
  • Meets ASTM F696-06 (2011) Standards
Not for electrical protection
Specifications
Certifications/ComplianceASTM F696-06 (2011)
DescriptionStraight thumb, Nylon pull strap
ColorNatural
HandSpecific
MaterialTop Grain Cowhide Leather
Product TypeGlove Protectors
Size10
For Use With (Application)Mechanical Protection
For Use With (Equipment)Class 1-2 Rubber Insulating Novax™ Gloves
Glove Cuff ColorGray
Glove Cuff StyleGauntlet
Hem ColorBrown
Series148-4000
Thumb StyleStraight
Length (English)12 in.
Length (English) Cuff3.6 in.
Length (Metric)30.5 cm
Length (Metric) Cuff9.1 cm
Unit SizeBOT