Benzyl Alcohol, 99%, Pure
Benzyl Alcohol, 99%, Pure
Benzyl Alcohol, 99%, Pure
Benzyl Alcohol, 99%, Pure
Thermo Scientific Chemicals
Benzyl Alcohol, 99%, Pure
CAS: 100-51-6 | C7H8O | 108.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 L
1 L
2.5 L
Packaging:
View all
Metal drum
Catalog number FSA148390250
Price (MYR)
3,496.00
EA
Add to cart
Quantity:
25 L
Packaging:
Metal drum
Request bulk or custom format
Price (MYR)
3,496.00
EA
Add to cart
Specifications
Chemical Name or MaterialBenzyl alcohol
CAS100-51-6
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0