4-Dimethylaminopyridine, 99%
4-Dimethylaminopyridine, 99%
4-Dimethylaminopyridine, 99%
4-Dimethylaminopyridine, 99%
Thermo Scientific Chemicals
4-Dimethylaminopyridine, 99%
CAS: 1122-58-3 | C7H10N2 | 122.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
25 g
100 g
Packaging:
View all
Glass bottle
Catalog number FSA148275000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material4-Dimethylaminopyridine
CAS1122-58-3
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0