3-Chloro-1,2-propanediol, 98%
3-Chloro-1,2-propanediol, 98%
3-Chloro-1,2-propanediol, 98%
3-Chloro-1,2-propanediol, 98%
Thermo Scientific Chemicals
3-Chloro-1,2-propanediol, 98%
CAS: 96-24-2 | C3H7ClO2 | 110.54 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
25 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number FSA148125000
Price (MYR)
638.00
BOT
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
638.00
BOT
Add to cart
Specifications
Chemical Name or Material3-Chloro-1, 2-propanediol
CAS96-24-2
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0