p-Toluoyl chloride, 98%
p-Toluoyl chloride, 98%
p-Toluoyl chloride, 98%
p-Toluoyl chloride, 98%
Thermo Scientific Chemicals
p-Toluoyl chloride, 98%
CAS: 874-60-2 | C8H7ClO | 154.6 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
Packaging:
Glass bottle
Catalog number FSA147980050
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Materialp-Toluoyl chloride
Name Note98%
CAS874-60-2
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement