1,1,2,2-Tetrachloroethane, 98.5%
1,1,2,2-Tetrachloroethane, 98.5%
1,1,2,2-Tetrachloroethane, 98.5%
1,1,2,2-Tetrachloroethane, 98.5%
Thermo Scientific Chemicals
1,1,2,2-Tetrachloroethane, 98.5%
CAS: 79-34-5 | C2H2Cl4 | 167.85 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
25 mL
1 L
Packaging:
Glass bottle
Catalog number FSA147941000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material1,1,2,2-Tetrachloroethane
Name Note98.5%
CAS79-34-5
CAS Min %98.0
Health Hazard 1GHS Signal Word: Danger