Formic acid, 98+%, pure
Formic acid, 98+%, pure
Formic acid, 98+%, pure
Formic acid, 98+%, pure
Thermo Scientific Chemicals
Formic acid, 98+%, pure
CAS: 64-18-6 | CH2O2 | 46.025 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
Packaging:
Plastic bottle
Catalog number FSA147932500
Price (MYR)
343.00
EA
Add to cart
Quantity:
250 mL
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
343.00
EA
Add to cart
Specifications
Chemical Name or MaterialFormic acid
CAS64-18-6
Health Hazard 1Danger
Assay Percent Range98+%
Molecular FormulaCH2O2