1,2-Bis(diphenylphosphino)ethane, 98+%
1,2-Bis(diphenylphosphino)ethane, 98+%
1,2-Bis(diphenylphosphino)ethane, 98+%
1,2-Bis(diphenylphosphino)ethane, 98+%
Thermo Scientific Chemicals
1,2-Bis(diphenylphosphino)ethane, 98+%
CAS: 1663-45-2 | C26H24P2 | 398.43 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
Packaging:
Glass bottle
Catalog number FSA147910010
Price (MYR)
282.00
BOT
Add to cart
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
282.00
BOT
Add to cart
Specifications
Chemical Name or Material1, 2-Bis(diphenylphosphino)ethane
Name Note98+%
CAS1663-45-2
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement: