p-Toluenesulfonamide, 99%
p-Toluenesulfonamide, 99%
p-Toluenesulfonamide, 99%
p-Toluenesulfonamide, 99%
Thermo Scientific Chemicals
p-Toluenesulfonamide, 99%
CAS: 70-55-3 | C7H9NO2S | 171.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Glass bottle
Catalog number FSA147692500
Price (MYR)
287.00
BOT
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
287.00
BOT
Add to cart
Specifications
Chemical Name or Materialp-Toluenesulfonamide
CAS70-55-3
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Harmful in contact with skin. May cause respiratory irritation. Harmful to aquatic life with long lasting effects. Causes skin irritation. Harmful if swallowed. Causes serious eye irritation. Harmful if inhaled. Contact with acids liberates very toxic gas.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection. Avoid breathing dust/fume/gas/mist/vapors/spray. IF ON SKIN: Wash with plenty of soap and water. Keep container tightly closed. IF ON SKIN: Wash with plenty of soap and water. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Avoid release to the environment. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. Wear protective gloves/protective clothing/eye protection/face protection.