1,1,1,3,3,3-Hexafluoro-2-propanol, 99.5+%, pure
1,1,1,3,3,3-Hexafluoro-2-propanol, 99.5+%, pure
1,1,1,3,3,3-Hexafluoro-2-propanol, 99.5+%, pure
1,1,1,3,3,3-Hexafluoro-2-propanol, 99.5+%, pure
Thermo Scientific Chemicals
1,1,1,3,3,3-Hexafluoro-2-propanol, 99.5+%, pure
CAS: 920-66-1 | C3H2F6O | 168.038 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
25 g
100 g
Packaging:
View all
Glass bottle
Catalog number FSA147545000
Price (MYR)
6,177.00
EA
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
6,177.00
EA
Add to cart
Specifications
Chemical Name or Material1,1,1,3,3,3-Hexafluoro-2-propanol
CAS920-66-1
CAS Min %99.5
Health Hazard 1Danger
CAS Max %100.0