4-Nitroanisole, 99+%
4-Nitroanisole, 99+%
4-Nitroanisole, 99+%
4-Nitroanisole, 99+%
Thermo Scientific Chemicals
4-Nitroanisole, 99+%
CAS: 100-17-4 | C7H7NO3 | 153.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA147020050
Price (MYR)
257.00
BOT
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
257.00
BOT
Add to cart
Specifications
Chemical Name or Material4-Nitroanisole
Name Note99+%
CAS100-17-4
CAS Min %99.0
Health Hazard 1GHS Signal Word: Warning