2,4-Diaminotoluene, 98%
2,4-Diaminotoluene, 98%
2,4-Diaminotoluene, 98%
2,4-Diaminotoluene, 98%
Thermo Scientific Chemicals
2,4-Diaminotoluene, 98%
CAS: 95-80-7 | C7H10N2 | 122.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
25 g
Packaging:
Glass bottle
Catalog number FSA146602500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material2, 4-Diaminotoluene
Name Note98%
CAS95-80-7
CAS Min %97.5
Health Hazard 1GHS Signal Word: Danger