Triphenylmethane, 99+%
Triphenylmethane, 99+%
Triphenylmethane, 99+%
Triphenylmethane, 99+%
Thermo Scientific Chemicals
Triphenylmethane, 99+%
CAS: 519-73-3 | C19H16 | 244.34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Packaging:
Glass bottle
Catalog number FSA146385000
Price (MYR)
2,276.00
EA
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,276.00
EA
Add to cart
Specifications
Chemical Name or MaterialTriphenylmethane
Name Note99+%
CAS519-73-3
Melting Point90.0°C to 94.0°C
ColorYellow