alpha-Bromo-2,3,4,5,6-pentafluorotoluene, 97%
alpha-Bromo-2,3,4,5,6-pentafluorotoluene, 97%
alpha-Bromo-2,3,4,5,6-pentafluorotoluene, 97%
alpha-Bromo-2,3,4,5,6-pentafluorotoluene, 97%
Thermo Scientific Chemicals
alpha-Bromo-2,3,4,5,6-pentafluorotoluene, 97%
CAS: 1765-40-8 | C7H2BrF5 | 260.98 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
5 g
10 g
Packaging:
Glass bottle
Catalog number FSA146330500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Materialα-Bromo-2,3,4,5,6-pentafluorotoluene
CAS1765-40-8
CAS Min %96.0
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0