4-Aminobenzoic Acid, 99%
4-Aminobenzoic Acid, 99%
4-Aminobenzoic Acid, 99%
4-Aminobenzoic Acid, 99%
Thermo Scientific Chemicals
4-Aminobenzoic Acid, 99%
CAS: 150-13-0 | C7H7NO2 | 137.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
Packaging:
View all
Glass bottle
Catalog number FSA146212500
Price (MYR)
336.00
EA
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
336.00
EA
Add to cart
Specifications
Chemical Name or Material4-Aminobenzoic acid
CAS150-13-0
CAS Min %98.5
Health Hazard 1Warning
CAS Max %100.0