3-Aminophenol, 99%
3-Aminophenol, 99%
3-Aminophenol, 99%
Thermo Scientific Chemicals
3-Aminophenol, 99%
CAS: 591-27-5 | C6H7NO | 109.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
Packaging:
Glass bottle
Catalog number FSA146102500
Price (MYR)
561.00
BOT
Add to cart
Quantity:
250 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
561.00
BOT
Add to cart
Specifications
Chemical Name or Material3-Aminophenol
Name Note99%
CAS591-27-5
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning