(Methoxymethyl)triphenylphosphonium chloride, 98%
(Methoxymethyl)triphenylphosphonium chloride, 98%
(Methoxymethyl)triphenylphosphonium chloride, 98%
(Methoxymethyl)triphenylphosphonium chloride, 98%
Thermo Scientific Chemicals
(Methoxymethyl)triphenylphosphonium chloride, 98%
CAS: 4009-98-7 | C20H20ClOP | 342.80 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
Packaging:
Glass bottle
Catalog number FSA146011000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material(Methoxymethyl)triphenylphosphonium chloride
Name Note98%
CAS4009-98-7
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
View more
For C-C Bond Formation (Olefination)
RUO – Research Use Only