m-Xylylenediamine, 99%
m-Xylylenediamine, 99%
m-Xylylenediamine, 99%
m-Xylylenediamine, 99%
Thermo Scientific Chemicals
m-Xylylenediamine, 99%
CAS: 1477-55-0 | C8H12N2 | 136.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
5 mL
100 mL
Packaging:
Glass bottle
Catalog number FSA141025000
Price (MYR)
1,142.00
EA
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,142.00
EA
Add to cart
Specifications
Chemical Name or Materialm-Xylylenediamine
CAS1477-55-0
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger
CAS Max %100.0