o-Xylene, 99%, pure
o-Xylene, 99%, pure
o-Xylene, 99%, pure
Thermo Scientific Chemicals
o-Xylene, 99%, pure
CAS: 95-47-6 | C8H10 | 106.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
1 L
Packaging:
Glass Bottle
Catalog number FSA140990025
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2.5 L
Packaging:
Glass Bottle
Request bulk or custom format
Specifications
Chemical Name or Materialo-Xylene
Melting Point-25.0°C
Name NotePure
CAS95-47-6