4-Vinylpyridine, 95%, stabilized
4-Vinylpyridine, 95%, stabilized
4-Vinylpyridine, 95%, stabilized
4-Vinylpyridine, 95%, stabilized
Thermo Scientific Chemicals
4-Vinylpyridine, 95%, stabilized
CAS: 100-43-6 | C7H7N | 105.14 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
100 mL
Packaging:
Glass bottle
Catalog number FSA140912500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material4-Vinylpyridine
Name Note95%
CAS100-43-6
CAS Min %94.0
Health Hazard 1GHS Signal Word: Danger