4-Vinyl-1-cyclohexene, 97%, stabilized
4-Vinyl-1-cyclohexene, 97%, stabilized
4-Vinyl-1-cyclohexene, 97%, stabilized
4-Vinyl-1-cyclohexene, 97%, stabilized
Thermo Scientific Chemicals
4-Vinyl-1-cyclohexene, 97%, stabilized
CAS: 100-40-3 | C8H12 | 108.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
Packaging:
Glass bottle
Catalog number FSA140885000
Price (MYR)
629.00
BOT
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
629.00
BOT
Add to cart
Specifications
Chemical Name or Material4-Vinyl-1-cyclohexene
Name Note99%
CAS100-40-3
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger