Vinyl acetate, 99+%, stabilized
Vinyl acetate, 99+%, stabilized
Vinyl acetate, 99+%, stabilized
Vinyl acetate, 99+%, stabilized
Thermo Scientific Chemicals
Vinyl acetate, 99+%, stabilized
CAS: 108-05-4 | C4H6O2 | 86.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number FSA140840250
Price (MYR)
292.00
BOT
Add to cart
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
292.00
BOT
Add to cart
Specifications
Chemical Name or MaterialVinyl acetate
CAS108-05-4
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Highly flammable liquid and vapor.
Harmful if inhaled.
CAS123-31-9