delta-Valerolactam, 99%
delta-Valerolactam, 99%
delta-Valerolactam, 99%
delta-Valerolactam, 99%
Thermo Scientific Chemicals
delta-Valerolactam, 99%
CAS: 675-20-7 | C5H9NO | 99.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA140780050
Price (MYR)
306.00
BOT
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
306.00
BOT
Add to cart
Specifications
Chemical Name or Materialδ-Valerolactam
Name Note99%
CAS675-20-7
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement: