Uridine, 99%
Uridine, 99%
Uridine, 99%
Thermo Scientific Chemicals
Uridine, 99%
CAS: 58-96-8 | C9H12N2O6 | 244.2 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
1 g
25 g
100 g
Packaging:
Glass bottle
Catalog number FSA140775000
Price (MYR)
7,237.00
EA
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
7,237.00
EA
Add to cart
Specifications
Chemical Name or MaterialUridine
CAS58-96-8
Absorbance0.72 to 0.76 (A250/A260), 0.34 to 0.38 (A280/A260)
Melting Point162.0°C to 170.0°C