2-Undecanone, 98%
2-Undecanone, 98%
2-Undecanone, 98%
Thermo Scientific Chemicals
2-Undecanone, 98%
CAS: 112-12-9 | C11H22O | 170.30 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
Packaging:
Glass bottle
Catalog number FSA140692500
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or Material2-Undecanone
Name Note99%
CAS112-12-9
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning