1-Undecanol, 98%
1-Undecanol, 98%
1-Undecanol, 98%
Thermo Scientific Chemicals
1-Undecanol, 98%
CAS: 112-42-5 | C11H24O | 172.31 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Packaging:
Glass bottle
Catalog number FSA140685000
Price (MYR)
851.00
BOT
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
851.00
BOT
Add to cart
Specifications
Chemical Name or Material1-Undecanol
Name Note98%
CAS112-42-5
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning