n-Undecane, 99%
n-Undecane, 99%
n-Undecane, 99%
Thermo Scientific Chemicals
n-Undecane, 99%
CAS: 1120-21-4 | C11H24 | 156.31 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
2.5 L
Catalog number FSA140665000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or Materialn-Undecane
Name Note99%
CAS1120-21-4
CAS Min %98.5
Health Hazard 1Danger