Tropolone, 98%
Tropolone, 98%
Tropolone, 98%
Thermo Scientific Chemicals
Tropolone, 98%
CAS: 533-75-5 | C7H6O2 | 122.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
Packaging:
Glass bottle
Catalog number FSA140580250
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialTropolone
CAS533-75-5
Melting Point50.0°C to 54.0°C
ColorBeige-Cream to White
EINECS Number208-577-2