Trithiocyanuric acid, 95%
Trithiocyanuric acid, 95%
Trithiocyanuric acid, 95%
Trithiocyanuric acid, 95%
Thermo Scientific Chemicals
Trithiocyanuric acid, 95%
CAS: 638-16-4 | C3H3N3S3 | 177.26 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA140530050
Price (MYR)
307.00
EA
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
307.00
EA
Add to cart
Specifications
Chemical Name or MaterialTrithiocyanuric acid
CAS638-16-4
Health Hazard 1GHS Signal Word: Warning
Health Hazard 2GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER