1,1,1-Tris(hydroxymethyl)ethane, 97%
1,1,1-Tris(hydroxymethyl)ethane, 97%
1,1,1-Tris(hydroxymethyl)ethane, 97%
1,1,1-Tris(hydroxymethyl)ethane, 97%
Thermo Scientific Chemicals
1,1,1-Tris(hydroxymethyl)ethane, 97%
CAS: 77-85-0 | C5H12O3 | 120.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
Packaging:
Plastic bottle
Catalog number FSA140515000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Specifications
Chemical Name or Material1, 1, 1-Tris(hydroxymethyl)ethane
Name Note97%
CAS77-85-0
CAS Min %96.0
Health Hazard 1GHS Signal Word: Warning