Trimethyl borate, 99%
Trimethyl borate, 99%
Trimethyl borate, 99%
Trimethyl borate, 99%
Trimethyl borate, 99%
Thermo Scientific Chemicals
Trimethyl borate, 99%
CAS: 121-43-7 | C3H9O3B | 103.91 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
Packaging:
Glass bottle
Catalog number FSA140125000
Price (MYR)
459.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
459.00
BOT
Add to cart
Specifications
Chemical Name or MaterialTrimethyl borate
Name Note99%
CAS121-43-7
Health Hazard 1Danger
Health Hazard 2GHS H Statement:
Harmful in contact with skin.
Highly flammable liquid and vapor.
View more
Suitable for battery materials development.
RUO – Research Use Only