3,4,5-Trimethoxyphenylacetic acid, 99%
3,4,5-Trimethoxyphenylacetic acid, 99%
3,4,5-Trimethoxyphenylacetic acid, 99%
3,4,5-Trimethoxyphenylacetic acid, 99%
Thermo Scientific Chemicals
3,4,5-Trimethoxyphenylacetic acid, 99%
CAS: 951-82-6 | C11H14O5 | 226.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
Packaging:
Glass bottle
Catalog number FSA139990050
Price (MYR)
364.00
BOT
Add to cart
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
364.00
BOT
Add to cart
Specifications
Chemical Name or Material3, 4, 5-Trimethoxyphenylacetic acid
CAS951-82-6
CAS Min %98.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0