alpha,alpha,alpha-Trifluorotoluene, 99+%
alpha,alpha,alpha-Trifluorotoluene, 99+%
alpha,alpha,alpha-Trifluorotoluene, 99+%
alpha,alpha,alpha-Trifluorotoluene, 99+%
Thermo Scientific Chemicals
alpha,alpha,alpha-Trifluorotoluene, 99+%
CAS: 98-08-8 | C7H5F3 | 146.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2.5 L
Packaging:
Plastic bottle
Catalog number FSA139770025
Price (MYR)
1,069.00
EA
Add to cart
Quantity:
2.5 L
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,069.00
EA
Add to cart
Specifications
Chemical Name or Materialα, α, α-Trifluorotoluene
Name Note99+%
CAS98-08-8
CAS Min %99.0
Health Hazard 1GHS Signal Word: Danger