2,2,2-Trifluoroethanol, 99.8%, extra pure
2,2,2-Trifluoroethanol, 99.8%, extra pure
2,2,2-Trifluoroethanol, 99.8%, extra pure
2,2,2-Trifluoroethanol, 99.8%, extra pure
Thermo Scientific Chemicals
2,2,2-Trifluoroethanol, 99.8%, extra pure
CAS: 75-89-8 | C2H3F3O | 100.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
25 mL
100 mL
2.5 L
Packaging:
Glass bottle
Catalog number FSA139755000
Price (MYR)
1,626.00
BOT
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,626.00
BOT
Add to cart
Specifications
Chemical Name or Material2, 2, 2-Trifluoroethanol
CAS75-89-8
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Flammable liquid and vapor.
Causes serious eye damage.
Toxic if swallowed.
Toxic if inhaled.
May damage fertility.
May cause damage to organs through prolonged or repeated exposure.
Health Hazard 3GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN:
View more