Triethylene glycol, 99%
Triethylene glycol, 99%
Triethylene glycol, 99%
Triethylene glycol, 99%
Thermo Scientific Chemicals
Triethylene glycol, 99%
CAS: 112-27-6 | C6H14O4 | 150.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 L
1 L
2.5 L
Packaging:
View all
Plastic drum
Catalog number FSA139590100
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 L
Packaging:
Plastic drum
Request bulk or custom format
Specifications
Chemical Name or MaterialTriethylene glycol
CAS112-27-6
Melting Point-7.0°C
ColorYellow
Density1.1200g/mL