2,4,6-Trichlorophenol, 98%
2,4,6-Trichlorophenol, 98%
2,4,6-Trichlorophenol, 98%
2,4,6-Trichlorophenol, 98%
Thermo Scientific Chemicals
2,4,6-Trichlorophenol, 98%
CAS: 88-06-2 | C6H3Cl3O | 197.45 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Glass bottle
Catalog number FSA139485000
Price (MYR)
887.00
EA
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
887.00
EA
Add to cart
Specifications
Chemical Name or Material2, 4, 6-Trichlorophenol
CAS88-06-2
CAS Min %97.5
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0