2,2,2-Trichloroethanol, 99%
2,2,2-Trichloroethanol, 99%
2,2,2-Trichloroethanol, 99%
2,2,2-Trichloroethanol, 99%
Thermo Scientific Chemicals
2,2,2-Trichloroethanol, 99%
CAS: 115-20-8 | C2H3Cl3O | 149.4 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Glass bottle
Catalog number FSA139445000
Price (MYR)
1,237.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,237.00
BOT
Add to cart
Specifications
Chemical Name or Material2, 2, 2-Trichloroethanol
Name Note99%
CAS115-20-8
CAS Min %98.5
Health Hazard 1GHS Signal Word: Danger