1,1,2-Trichloroethane, 98%
1,1,2-Trichloroethane, 98%
1,1,2-Trichloroethane, 98%
1,1,2-Trichloroethane, 98%
Thermo Scientific Chemicals
1,1,2-Trichloroethane, 98%
CAS: 79-00-5 | C2H3Cl3 | 133.40 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mL
1 L
2.5 L
Packaging:
Glass bottle
Catalog number FSA139432500
Price (MYR)
578.00
BOT
Add to cart
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
578.00
BOT
Add to cart
Specifications
Chemical Name or Material1, 1, 2-Trichloroethane
CAS79-00-5
Health Hazard 1GHS Signal Word: Warning
Melting Point-37.0°C