Trichloroacetonitrile, 98%
Trichloroacetonitrile, 98%
Trichloroacetonitrile, 98%
Trichloroacetonitrile, 98%
Thermo Scientific Chemicals
Trichloroacetonitrile, 98%
CAS: 545-06-2 | C2Cl3N | 144.39 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
5 g
100 g
Packaging:
Glass bottle
Catalog number FSA139375000
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Specifications
Chemical Name or MaterialTrichloroacetonitrile
CAS545-06-2
Health Hazard 1GHS Signal Word: Danger
Melting Point-42.0°C
ColorColorless to Yellow