Tri-n-butylphosphine, 95%
Tri-n-butylphosphine, 95%
Tri-n-butylphosphine, 95%
Tri-n-butylphosphine, 95%
Thermo Scientific Chemicals
Tri-n-butylphosphine, 95%
CAS: 998-40-3 | C12H27P | 202.32 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Packaging:
Glass bottle
Catalog number FSA139345000
Price (MYR)
2,694.00
BOT
Add to cart
Quantity:
500 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,694.00
BOT
Add to cart
Specifications
Chemical Name or MaterialTri-n-butylphosphine
Name Note95%
CAS998-40-3
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Flammable liquid and vapor.
Catches fire spontaneously if exposed to air.
Harmful if swallowed.
Harmful in contact with skin.
Causes severe skin burns and eye damage.
Toxic to aquatic life with long la