1,2,4-1H-Triazole, 99.5%
1,2,4-1H-Triazole, 99.5%
1,2,4-1H-Triazole, 99.5%
1,2,4-1H-Triazole, 99.5%
Thermo Scientific Chemicals
1,2,4-1H-Triazole, 99.5%
CAS: 288-88-0 | C2H3N3 | 69.06 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
View all
Plastic bottle
Catalog number FSA139280250
Price (MYR)
255.00
BOT
Add to cart
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
255.00
BOT
Add to cart
Specifications
Chemical Name or Material1, 2, 4-1H-Triazole
CAS288-88-0
CAS Min %99.4
Health Hazard 1GHS Signal Word: Warning
CAS Max %100.0