2,4,6-Triaminopyrimidine, 97%
2,4,6-Triaminopyrimidine, 97%
2,4,6-Triaminopyrimidine, 97%
2,4,6-Triaminopyrimidine, 97%
Thermo Scientific Chemicals
2,4,6-Triaminopyrimidine, 97%
CAS: 1004-38-2 | C4H7N5 | 125.135 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Plastic bottle
Catalog number FSA139251000
Price (MYR)
1,311.00
BOT
Add to cart
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,311.00
BOT
Add to cart
Specifications
Chemical Name or Material2, 4, 6-Triaminopyrimidine
Name Note97%
CAS1004-38-2
CAS Min %96.0
CAS Max %100.0