Tri-O-acetyl-D-glucal, 99%
Tri-O-acetyl-D-glucal, 99%
Tri-O-acetyl-D-glucal, 99%
Tri-O-acetyl-D-glucal, 99%
Thermo Scientific Chemicals
Tri-O-acetyl-D-glucal, 99%
CAS: 2873-29-2 | C12H16O7 | 272.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Glass bottle
Catalog number FSA139241000
Price (MYR)
2,396.00
EA
Add to cart
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
2,396.00
EA
Add to cart
Specifications
Chemical Name or MaterialTri-O-acetyl-D-glucal
Name Note99%
CAS2873-29-2
Melting Point51.0°C to 53.0°C