Thiosemicarbazide, 98+%
Thiosemicarbazide, 98+%
Thiosemicarbazide, 98+%
Thiosemicarbazide, 98+%
Thermo Scientific Chemicals
Thiosemicarbazide, 98+%
CAS: 79-19-6 | CH5N3S | 91.13 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Plastic bottle
Catalog number FSA138905000
Price (MYR)
1,063.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (MYR)
1,063.00
BOT
Add to cart
Specifications
Chemical Name or MaterialThiosemicarbazide
Name Note99%
CAS79-19-6
Health Hazard 1GHS Signal Word: Danger
Health Hazard 2GHS H Statement
Fatal if swallowed.