Thiosalicylic acid, 98%
Thiosalicylic acid, 98%
Thiosalicylic acid, 98%
Thiosalicylic acid, 98%
Thermo Scientific Chemicals
Thiosalicylic acid, 98%
CAS: 147-93-3 | C7H6O2S | 154.183 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Packaging:
Glass bottle
Catalog number FSA138895000
Price (MYR)
1,550.00
BOT
Add to cart
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Price (MYR)
1,550.00
BOT
Add to cart
Specifications
Chemical Name or MaterialThiosalicylic acid
Name Note98%
CAS147-93-3
Melting Point164.0°C to 169.0°C
Health Hazard 1GHS Signal Word: Warning